Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198368.1 | NP_938182.1 | 5 - 6 | 605 | 123 | |
XM_009292430.3 | XP_009290705.1 | 5 - 6 | 598 | 123 | ||
GenBank mRNA | BC050160.1 | - | 5 - 6 | 631 | 123 | |
BC056696.1 | - | 5 - 6 | 605 | 123 | ||
BC065878.1 | - | 5 - 6 | 585 | 123 | ||
BC152198.1 | - | 5 - 6 | 635 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |