Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199773.1 | NP_956067.1 | 2 - 3 | 891 | 133 | |
GenBank mRNA | BC045359.1 | - | 2 - 3 | 891 | 133 | |
BC164141.1 | - | 2 - 3 | 808 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |