Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stard14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200220.1 | NP_956514.1 | 5 - 6 | 747 | 86 | |
XM_005173231.3 | XP_005173288.1 | 6 - 7 | 719 | 86 | ||
GenBank mRNA | BC046038.1 | - | 5 - 6 | 747 | 86 | |
BC063977.1 | - | 5 - 6 | 659 | 86 | ||
BC171583.1 | - | 5 - 6 | 675 | 86 | ||
BC171587.1 | - | 5 - 6 | 675 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |