Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pias4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242942.1 | NP_001229871.1 | 7 - 8 | 856 | 94 | |
NM_200343.2 | NP_956637.2 | 7 - 8 | 912 | 94 | ||
GenBank mRNA | BC053149.1 | - | 7 - 8 | 896 | 94 | |
EH570193.1 | - | 4 - 5 | 753 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |