Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200645.1 | NP_956939.1 | 3 - 4 | 1052 | 84 | |
GenBank mRNA | BC044529.1 | - | 1 - 2 | 1854 | 84 | |
BC057464.1 | - | 3 - 4 | 1052 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |