Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esama | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115066.1 | NP_001108538.1 | 6 - 7 | 898 | 82 | |
GenBank mRNA | CU638747.1 | - | 6 - 7 | 898 | 82 | |
FJ205704.1 | - | 6 - 7 | 875 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Probe spans exons |