Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoxb7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115091.2 | NP_001108563.1 | 1 - 2 | 486 | 72 | |
GenBank mRNA | BC163357.1 | - | 1 - 2 | 486 | 72 | |
BC163361.1 | - | 1 - 2 | 486 | 72 | ||
DQ060542.1 | - | 1 - 2 | 277 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |