Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115133.1 | NP_001108605.1 | 4 - 5 | 493 | 71 | |
GenBank mRNA | BC056715.1 | - | 4 - 5 | 464 | 71 | |
DR722002.1 | - | 4 - 5 | 512 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |