Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpeb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351720.1 | NP_001338649.1 | 5 - 6 | 1604 | 76 | |
NM_200981.2 | NP_957275.2 | 5 - 6 | 1556 | 76 | ||
XM_005173113.4 | XP_005173170.2 | 6 - 7 | 1656 | 76 | ||
XM_009291021.3 | XP_009289296.1 | 6 - 7 | 1986 | 76 | ||
GenBank mRNA | BC055522.1 | - | 5 - 6 | 1556 | 76 | |
BC165168.1 | - | 4 - 5 | 1299 | 76 | ||
EF033337.1 | - | 4 - 5 | 1751 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |