Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtnbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201134.1 | NP_957428.1 | 1 - 2 | 119 | 58 | |
XM_005158097.3 | XP_005158154.1 | 1 - 2 | 386 | 58 | ||
XM_005158098.4 | XP_005158155.1 | 1 - 2 | 387 | 58 | ||
XM_005158099.4 | XP_005158156.1 | 1 - 2 | 386 | 58 | ||
XM_005158100.3 | XP_005158157.1 | 1 - 2 | 387 | 58 | ||
XM_005158101.3 | XP_005158158.1 | 1 - 2 | 386 | 58 | ||
XM_021466652.1 | XP_021322327.1 | 1 - 2 | 389 | 58 | ||
GenBank mRNA | BC049451.1 | - | 1 - 2 | 119 | 58 | |
BC152672.1 | - | 1 - 2 | 357 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |