Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cct6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201290.1 | NP_958447.1 | 4 - 5 | 548 | 72 | |
XM_005161447.2 | XP_005161504.1 | 5 - 6 | 902 | 72 | ||
GenBank mRNA | BC044393.1 | - | 4 - 5 | 548 | 72 | |
BC071416.1 | - | 4 - 5 | 585 | 72 | ||
BC155176.1 | - | 5 - 6 | 909 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |