Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cct6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201290.1 | NP_958447.1 | 5 - 6 | 654 | 91 | |
XM_005161447.2 | XP_005161504.1 | 6 - 7 | 1008 | 91 | ||
GenBank mRNA | BC044393.1 | - | 5 - 6 | 654 | 91 | |
BC071416.1 | - | 5 - 6 | 691 | 91 | ||
BC155176.1 | - | 6 - 7 | 1015 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |