Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec22bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328526.1 | NP_001315455.1 | 3 - 4 | 327 | 74 | |
NM_201302.2 | NP_958459.1 | 2 - 3 | 315 | 74 | ||
GenBank mRNA | BC055515.1 | - | 2 - 3 | 271 | 74 | |
BC152220.1 | - | 3 - 4 | 314 | 74 | ||
EH489983.1 | - | 2 - 3 | 304 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |