Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205658.1 | NP_991221.1 | 714 | 85 | ||
GenBank mRNA | BC065880.1 | - | 714 | 85 |
Gene Symbol: LOC110438456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021470923.1 | XP_021326598.1 | 1 - 2 | 180 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |