Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdpk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205699.1 | 9 - 10 | 943 | 74 | ||
GenBank mRNA | BC065959.1 | - | 9 - 10 | 943 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |