Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207083.1 | NP_996966.1 | 3 - 4 | 328 | 80 | |
GenBank mRNA | BC066472.1 | - | 3 - 4 | 328 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |