Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttn.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021479071.1 | XP_021334746.1 | 183 - 184 | 66195 | 112 | |
XM_021479072.1 | XP_021334747.1 | 141 - 142 | 58594 | 112 | ||
GenBank mRNA | CK676433.1 | - | 2 - 3 | 459 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |