Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328059.1 | NP_001314988.1 | 13 - 14 | 1483 | 76 | |
GenBank mRNA | BC057475.1 | - | 13 - 14 | 1454 | 76 | |
BC154320.1 | - | 13 - 14 | 1447 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |