Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrpkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212991.3 | NP_998156.3 | 13 - 14 | 1139 | 105 | |
GenBank mRNA | BC096867.1 | - | 13 - 14 | 1119 | 105 | |
BC164732.1 | - | 12 - 13 | 1061 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |