Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif3s6ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213128.1 | NP_998293.1 | 3 - 4 | 373 | 96 | |
GenBank mRNA | BC054626.1 | - | 3 - 4 | 373 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |