Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: khdrbs1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213235.1 | NP_998400.1 | 6 - 7 | 974 | 75 | |
GenBank mRNA | BC067711.1 | - | 6 - 7 | 974 | 75 | |
BC155179.1 | - | 6 - 7 | 995 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |