Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213311.2 | NP_998476.1 | 4 - 5 | 702 | 63 | |
GenBank mRNA | BC066610.1 | - | 4 - 5 | 63 | ||
EB919906.1 | - | 1 - 2 | 87 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |