Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214733.2 | NP_999898.2 | 3 - 4 | 483 | 94 | |
GenBank mRNA | BC065984.1 | - | 3 - 4 | 450 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |