Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hs3st3b1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080569.1 | NP_001074038.1 | 2 | 851 | 128 | |
GenBank mRNA | DQ812987.1 | - | 2 | 851 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |