Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zic6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001837.1 | NP_001001837.1 | 2 | 1455 | 141 | |
GenBank mRNA | AY572956.1 | - | 2 | 1455 | 141 | |
BC162343.1 | - | 2 | 1450 | 141 | ||
BC162640.1 | - | 2 | 1450 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Inventoried Both primers and probe map within a single exon |