Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131273.1 | NP_571348.1 | 1 | 256 | 87 | |
GenBank mRNA | BC095601.1 | - | 1 | 284 | 113 | |
BC162283.1 | - | 1 | 309 | 87 | ||
BC162290.1 | - | 1 | 309 | 87 | ||
U50563.1 | - | 1 | 256 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Amplicon spans exons and probe does not span exons |