Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081873.1 | NP_001075342.1 | 2 - 3 | 297 | 73 | |
XM_005603492.3 | XP_005603549.1 | 291 | 73 | |||
GenBank mRNA | AJ315379.1 | - | 2 - 3 | 297 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Equus caballus Probe spans exons |