Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081893.1 | NP_001075362.1 | 4 - 5 | 679 | 65 | |
GenBank mRNA | AJ428063.1 | - | 2 - 3 | 449 | 65 | |
AY290704.1 | - | 4 - 5 | 679 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |