Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILT11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082527.1 | NP_001075996.1 | 4 - 5 | 727 | 95 | |
XM_005596095.2 | XP_005596152.1 | 710 | 95 | |||
GenBank mRNA | AB120409.1 | - | 4 - 5 | 727 | 95 | |
AB120410.1 | - | 3 - 4 | 664 | 95 | ||
AB120411.1 | - | 4 - 5 | 682 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Equus caballus Probe spans exons |