Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081782.2 | NP_001075251.1 | 1 | 652 | 91 | |
GenBank mRNA | AB264325.2 | - | 1 | 117 | 91 | |
AB267709.1 | - | 1 | 652 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Equus caballus Both primers and probe map within a single exon |