Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163887.1 | NP_001157359.1 | 2 - 3 | 190 | 82 | |
GenBank mRNA | AJ251191.1 | - | 2 - 3 | 175 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Equus caballus Probe spans exons |