Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195221.1 | NP_001182150.1 | 2 - 3 | 333 | 130 | |
GenBank mRNA | AJ319906.1 | - | 2 - 3 | 161 | 130 | |
HM769759.1 | - | 2 - 3 | 333 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Equus caballus Probe spans exons |