Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501733.4 | XP_001501783.1 | 559 | 78 | ||
GenBank mRNA | AY246738.1 | - | 2 - 3 | 262 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Equus caballus Probe spans exons |