Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163953.1 | NP_001157425.1 | 5 - 6 | 943 | 66 | |
XM_014728733.2 | XP_014584219.1 | 1918 | 66 | |||
XM_023633178.1 | XP_023488946.1 | 1277 | 66 | |||
XM_023633179.1 | XP_023488947.1 | 3353 | 66 | |||
GenBank mRNA | DQ367333.1 | - | 2 - 3 | 189 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |