Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242444.1 | NP_001229373.1 | 1 | 48 | 72 | |
XM_014735597.1 | XP_014591083.1 | 141 | 72 | |||
GenBank mRNA | EU810393.1 | - | 1 | 48 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Both primers and probe map within a single exon |