Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159692.1 | NP_001153164.1 | 2 - 3 | 342 | 76 | |
XM_023640254.1 | XP_023496022.1 | 515 | 76 | |||
GenBank mRNA | FJ028654.1 | - | 2 - 3 | 342 | 76 | |
JF749865.1 | - | 2 - 3 | 326 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Equus caballus Probe spans exons |