Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205192.1 | NP_001192121.1 | 7 - 8 | 1050 | 98 | |
XM_023644173.1 | XP_023499941.1 | 1591 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Equus caballus Probe spans exons |