Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFA5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166097.1 | NP_001159569.1 | 1 - 2 | 177 | 114 | |
GenBank mRNA | GQ259801.1 | - | 1 - 2 | 177 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Equus caballus Probe spans exons |