Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168391.1 | NP_001161863.1 | 4 - 5 | 462 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |