Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170767.2 | NP_001164238.1 | 3 - 4 | 130 | 91 | |
XM_014738515.2 | XP_014594001.1 | 90 | 91 | |||
GenBank mRNA | GU196267.1 | - | 3 - 4 | 130 | 91 | |
KP658381.1 | - | 2 - 3 | 86 | 91 | ||
KP658382.1 | - | 2 - 3 | 86 | 91 | ||
KT368778.1 | - | 2 - 3 | 80 | 91 | ||
KT368779.1 | - | 2 - 3 | 80 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Equus caballus Probe spans exons |