Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELPLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105161.1 | NP_001098631.1 | 2 | 921 | 119 | |
XM_005612335.3 | XP_005612392.2 | 1196 | 119 | |||
GenBank mRNA | AM778466.2 | - | 2 | 921 | 119 | |
AY298766.1 | - | 2 | 903 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon |