Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305651.2 | NP_001292580.2 | 10 - 11 | 1542 | 104 | |
XM_015283458.2 | XP_015138944.1 | 10 - 11 | 1492 | 104 | ||
GenBank mRNA | BX929305.1 | - | 1 - 2 | 155 | 104 | |
BX934454.1 | - | 3 - 4 | 490 | 104 | ||
CR407217.1 | - | 3 - 4 | 380 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |