Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417405.6 | XP_417405.4 | 3 - 4 | 184 | 67 | |
GenBank mRNA | BX932886.1 | - | 2 - 3 | 152 | 67 | |
BX950317.2 | - | 2 - 3 | 148 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |