Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198602.1 | NP_001185531.1 | 4 - 5 | 328 | 100 | |
XM_015278878.2 | XP_015134364.1 | 4 - 5 | 349 | 100 | ||
GenBank mRNA | BX933615.1 | - | 4 - 5 | 345 | 100 | |
BX933650.2 | - | 4 - 5 | 345 | 100 | ||
BX933652.1 | - | 4 - 5 | 345 | 100 | ||
BX936227.2 | - | 4 - 5 | 345 | 100 | ||
BX936267.2 | - | 4 - 5 | 345 | 100 | ||
CR353143.1 | - | 4 - 5 | 333 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |