Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195594.1 | NP_001182523.1 | 2 - 3 | 262 | 109 | |
GenBank mRNA | BX931314.2 | - | 2 - 3 | 251 | 109 | |
BX934303.1 | - | 2 - 3 | 254 | 109 | ||
BX934376.1 | - | 2 - 3 | 254 | 109 | ||
BX935522.1 | - | 2 - 3 | 262 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |