Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277465.1 | NP_001264394.1 | 2 - 3 | 195 | 74 | |
GenBank mRNA | BX931358.2 | - | 2 - 3 | 187 | 74 | |
BX933533.2 | - | 2 - 3 | 161 | 74 | ||
CF256811.1 | - | 2 - 3 | 197 | 74 | ||
CR386803.1 | - | 2 - 3 | 173 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |