Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418224.6 | XP_418224.2 | 2 - 3 | 268 | 61 | |
GenBank mRNA | BX929663.1 | - | 2 - 3 | 252 | 61 | |
BX930905.2 | - | 2 - 3 | 251 | 61 | ||
BX935423.2 | - | 2 - 3 | 255 | 61 | ||
BX935442.2 | - | 2 - 3 | 243 | 61 | ||
CR385484.1 | - | 2 - 3 | 231 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |