Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277613.1 | NP_001264542.1 | 4 - 5 | 393 | 102 | |
GenBank mRNA | BU330715.1 | - | 4 - 5 | 393 | 102 | |
BX933248.1 | - | 4 - 5 | 377 | 102 | ||
BX935707.2 | - | 4 - 5 | 336 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |