Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSKH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277682.1 | NP_001264611.1 | 2 - 3 | 1147 | 78 | |
XM_015279128.2 | XP_015134614.1 | 3 - 4 | 1235 | 78 | ||
GenBank mRNA | BX935768.2 | - | 2 - 3 | 1147 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |