Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416415.6 | XP_416415.5 | 2 - 3 | 961 | 77 | |
GenBank mRNA | CR353321.1 | - | 2 - 3 | 276 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |